3-Methyl-2-oxo-2,3-dihydro-1H-imidazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

059590-500MG
101918-580EA 855.38 USD
101918-580
3-Methyl-2-oxo-2,3-dihydro-1H-imidazole-4-carboxylic acid ≥95%
3-Methyl-2-oxo-2,3-dihydro-1H-imidazole-4-carboxylic acid

Hazard: Irritant

Formula: C₅H₆N₂O₃
Melting Pt: 222 °C
MDL Number: MFCD14586338
CAS Number: 17245-60-2

Order Now

Specification Test Results

Melting point 222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR