2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine ≥95%

Supplier: Matrix Scientific

059556-500MG
101918-420EA 526.15 USD
101918-420
2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine ≥95%
2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine

Hazard: Irritant

Formula: C₇H₆Cl₂N₂
MW: 189.0419 g/mol
Melting Pt: 77…79 °C
MDL Number: MFCD09750204
CAS Number: 5466-43-3

Order Now

Specification Test Results

Melting point 77-79°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR