6-[(4-Methoxyphenyl)sulfonyl]-3-pyridinylamine ≥95%
Supplier: Matrix Scientific
054893-1G
054893-500MG
101908-954EA
927.69
USD
101908-954
101908-952
6-[(4-Methoxyphenyl)sulfonyl]-3-pyridinylamine ≥95%
6-[(4-Methoxyphenyl)sulfonyl]-3-pyridinylamine
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₃S MW: 264.31 g/mol Melting Pt: 180…182 °C |
MDL Number:
MFCD16140353 CAS Number: 1221792-06-8 |
Specification Test Results
Melting point | 180-182°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...