6-[(4-Methoxyphenyl)sulfanyl]-3-pyridinylamine ≥95%
Supplier: Matrix Scientific
054892-1G
054892-500MG
101908-950EA
698.46
USD
101908-950
101908-948
6-[(4-Methoxyphenyl)sulfanyl]-3-pyridinylamine ≥95%
6-[(4-Methoxyphenyl)sulfanyl]-3-pyridinylamine
Hazard: Irritant
Formula:
C₁₂H₁₂N₂OS MW: 232.31 g/mol Melting Pt: 67…69 °C |
MDL Number:
MFCD11132352 CAS Number: 1019363-45-1 |
Specification Test Results
Melting point | 67-69°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...