8-Cyclohexyl-1-thia-4,8-diazaspiro[4.5]decane hydrochloride ≥95%

Supplier: Matrix Scientific

054880-500MG
101908-896EA 556.92 USD
101908-896
8-Cyclohexyl-1-thia-4,8-diazaspiro[4.5]decane hydrochloride ≥95%
8-Cyclohexyl-1-thia-4,8-diazaspiro[4.5]decane hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₅ClN₂S
Melting Pt: 171…172 °C
MDL Number: MFCD16140344
CAS Number: 1221791-74-7

Order Now

Specification Test Results

Melting point 171-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR