3-Cyano-6-cyclopropyl-2-pyridine carboxylic acid ≥95%
Supplier: Matrix Scientific
054878-1G
054878-500MG
101908-890EA
838.46
USD
101908-890
101908-888
3-Cyano-6-cyclopropyl-2-pyridine carboxylic acid ≥95%
3-Cyano-6-cyclopropyl-2-pyridine carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₈N₂O₂ MW: 188.19 g/mol Melting Pt: 155 °C (dec.) |
MDL Number:
MFCD16140342 CAS Number: 1221791-98-5 |
Specification Test Results
Melting point | 155°C(dec) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...