3-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
Supplier: Matrix Scientific
054874-1G
054874-500MG
101908-874EA
356.92
USD
101908-874
101908-876
3-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
3-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Hazard: Irritant
Formula:
C₈H₄ClF₃N₂ Melting Pt: 41…44 °C |
MDL Number:
MFCD09994776 CAS Number: 1019027-76-9 |
Specification Test Results
Melting point | 41-44°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...