6-[(4-Bromophenyl)sulfinyl]-3-pyridinylamine ≥95%

Supplier: Matrix Scientific

054871-500MG
101908-866EA 921.54 USD
101908-866
6-[(4-Bromophenyl)sulfinyl]-3-pyridinylamine ≥95%
6-[(4-Bromophenyl)sulfinyl]-3-pyridinylamine

Hazard: Irritant

Formula: C₁₁H₉BrN₂OS
MW: 297.18 g/mol
Melting Pt: 168…170 °C
MDL Number: MFCD16140338
CAS Number: 1221791-64-5

Order Now

Specification Test Results

Melting point 168-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR