5-(4-Bromo-3-fluorophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione ≥95%
Supplier: Matrix Scientific
054866-1G
054866-500MG
101908-850EA
467.69
USD
101908-850
101908-848
5-(4-Bromo-3-fluorophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione ≥95%
5-(4-Bromo-3-fluorophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione
Hazard: Irritant
Formula:
C₉H₇BrFN₃S Melting Pt: 251…253 °C |
MDL Number:
MFCD16140336 CAS Number: 1221792-04-6 |
Specification Test Results
Melting point | 251-253°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...