2-(Azetidin-3-yl)pyridine dihydrochloride ≥95%

Supplier: Matrix Scientific

054861-500MG 054861-1G
101908-826EA 1163.08 USD
101908-826 101908-824
2-(Azetidin-3-yl)pyridine dihydrochloride ≥95%
2-(Azetidin-3-yl)pyridine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₂Cl₂N₂
Melting Pt: 174…175 °C
MDL Number: MFCD16140331
CAS Number: 206446-45-9

Order Now

Specification Test Results

Melting point 174-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR