8-Amino-6,7-diazaspiro[3.4]oct-7-en-5-one ≥95%

Supplier: Matrix Scientific

054858-1G 054858-500MG
101908-816EA 1163.08 USD
101908-816 101908-814
8-Amino-6,7-diazaspiro[3.4]oct-7-en-5-one ≥95%
8-Amino-6,7-diazaspiro[3.4]oct-7-en-5-one

Hazard: Irritant

Formula: C₆H₉N₃O
Melting Pt: 86…88 °C
MDL Number: MFCD16140329
CAS Number: 1221791-84-9

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR