5-(Trifluoromethyl)-1H-indole-7-carbonitrile ≥95%

Supplier: Matrix Scientific

054856-500MG 054856-1G
101908-808EA 1109.23 USD
101908-808 101908-810
5-(Trifluoromethyl)-1H-indole-7-carbonitrile ≥95%
5-(Trifluoromethyl)-1H-indole-7-carbonitrile

Hazard: Irritant

Formula: C₁₀H₅F₃N₂
Melting Pt: 151…152 °C
MDL Number: MFCD16140327
CAS Number: 1221792-90-0

Order Now

Specification Test Results

Melting point 151-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR