1-Thia-4-azaspiro[4.6]undecane hydrochloride ≥95%

Supplier: Matrix Scientific

054850-500MG
101908-784EA 375.38 USD
101908-784
1-Thia-4-azaspiro[4.6]undecane hydrochloride ≥95%
1-Thia-4-azaspiro[4.6]undecane hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNS
Melting Pt: 157…159 °C
MDL Number: MFCD16140325
CAS Number: 875-11-6

Order Now

Specification Test Results

Melting point 157-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR