6-(Phenylsulfanyl)-3-pyridinamine ≥95%

Supplier: Matrix Scientific

054826-500MG 054826-1G
101908-688EA 556.92 USD
101908-688 101908-690
6-(Phenylsulfanyl)-3-pyridinamine ≥95%
6-(Phenylsulfanyl)-3-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₀N₂S
MW: 202.28 g/mol
Melting Pt: 118…120 °C
MDL Number: MFCD03230019
CAS Number: 103983-07-9

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR