4-Methyl-5,6,7,8-tetrahydro-2(1H)-quinolinone ≥95%

Supplier: Matrix Scientific

054811-500MG
101908-628EA 356.92 USD
101908-628
4-Methyl-5,6,7,8-tetrahydro-2(1H)-quinolinone ≥95%
4-Methyl-5,6,7,8-tetrahydro-2(1H)-quinolinone

Hazard: Irritant

Formula: C₁₀H₁₃NO
MW: 163.22 g/mol
Melting Pt: 252…254 °C
MDL Number: MFCD04037462
CAS Number: 16236-70-7

Order Now

Specification Test Results

Melting point 252-254°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR