Methyl-5-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-7-carboxylate ≥95%

Supplier: Matrix Scientific

054799-500MG 054799-1G
101908-578EA 529.23 USD
101908-578 101908-580
Methyl-5-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-7-carboxylate ≥95%
Methyl-5-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-7-carboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₄
Melting Pt: 234…236 °C
MDL Number: MFCD16140305
CAS Number: 1221792-45-5

Order Now

Specification Test Results

Melting point 234-236°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR