1-[2-(Methoxycarbonyl)-4-nitrophenyl]-3-azetanecarboxylic acid ≥95%

Supplier: Matrix Scientific

054781-500MG 054781-1G
101908-504EA 556.92 USD
101908-504 101908-506
1-[2-(Methoxycarbonyl)-4-nitrophenyl]-3-azetanecarboxylic acid ≥95%
1-[2-(Methoxycarbonyl)-4-nitrophenyl]-3-azetanecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₆
MW: 280.24 g/mol
Melting Pt: 178…180 °C
MDL Number: MFCD16140290
CAS Number: 1234873-83-6

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR