Ethyl-3-oxo-1-thia-4-azaspiro[4.5]decane-6-carboxylate ≥95%

Supplier: Matrix Scientific

054747-500MG
101908-372EA 356.92 USD
101908-372
Ethyl-3-oxo-1-thia-4-azaspiro[4.5]decane-6-carboxylate ≥95%
Ethyl-3-oxo-1-thia-4-azaspiro[4.5]decane-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₇NO₃S
Melting Pt: 126…128 °C
MDL Number: MFCD16140286
CAS Number: 1194701-70-6

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR