Ethyl-5-acetyl-2-pyridinecarboxylate ≥97%

Supplier: Matrix Scientific

054733-1G 054733-500MG
101908-316EA 823.08 USD
101908-316 101908-314
Ethyl-5-acetyl-2-pyridinecarboxylate ≥97%
Ethyl-5-acetyl-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MW: 193.2 g/mol
Melting Pt: 73…75 °C
MDL Number: MFCD16140282
CAS Number: 99060-45-4

Order Now

Specification Test Results

Melting point 73-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR