Diethyl-3-cyano-6-cyclopropyl-2,5-pyridinedicarboxylate ≥95%

Supplier: Matrix Scientific

054713-500MG
101908-238EA 1109.23 USD
101908-238
Diethyl-3-cyano-6-cyclopropyl-2,5-pyridinedicarboxylate ≥95%
Diethyl-3-cyano-6-cyclopropyl-2,5-pyridinedicarboxylate

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₄
MW: 288.31 g/mol
Melting Pt: 117…119 °C
MDL Number: MFCD16140279
CAS Number: 1221792-74-0

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR