2-(Diethylamino)-4-pyrimidinecarbonitrile ≥95%

Supplier: Matrix Scientific

054711-500MG
101908-232EA 747.69 USD
101908-232
2-(Diethylamino)-4-pyrimidinecarbonitrile ≥95%
2-(Diethylamino)-4-pyrimidinecarbonitrile

Hazard: Irritant

Formula: C₉H₁₂N₄
MW: 176.22 g/mol
Melting Pt: 50…51 °C
MDL Number: MFCD15202259
CAS Number: 75825-49-9

Order Now

Specification Test Results

Melting point 50-51°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR