2-Amino-3-iodo-5-(trifluoromethyl)benzenecarbonitrile ≥95%
Supplier: Matrix Scientific
054673-500MG
054673-1G
101908-078EA
698.46
USD
101908-078
101908-076
2-Amino-3-iodo-5-(trifluoromethyl)benzenecarbonitrile ≥95%
2-Amino-3-iodo-5-(trifluoromethyl)benzenecarbonitrile
Hazard: Irritant
Formula:
C₈H₄F₃IN₂ MW: 312.03 g/mol Melting Pt: 124…126 °C |
MDL Number:
MFCD16140261 CAS Number: 1221792-71-7 |
Specification Test Results
Melting point | 124-126°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...