3-Acetyl-2-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Acetyl-2-pyridinecarboxylic acid

054663-500MG 054663-1G
101908-038EA 700 USD
101908-038 101908-040
3-Acetyl-2-pyridine carboxylic acid ≥95%
3-Acetyl-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Melting Pt: 131 °C (dec.)
MDL Number: MFCD16140257
CAS Number: 716362-04-8

Order Now

Specification Test Results

Melting point 131dec°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR