6-Bromo-2-chloro-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

054564-500MG
101907-780EA 1109.23 USD
101907-780
6-Bromo-2-chloro-1H-indole-3-carbaldehyde ≥95%
6-Bromo-2-chloro-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₉H₅BrClNO
MW: 258.51 g/mol
Melting Pt: 257…258 °C
MDL Number: MFCD01415811
CAS Number: 1221792-00-2

Order Now

Specification Test Results

Melting point 257-258°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR