6-Bromo-2-(dimethylamino)-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

054560-500MG
101907-768EA 556.92 USD
101907-768
6-Bromo-2-(dimethylamino)-1H-indole-3-carbaldehyde ≥95%
6-Bromo-2-(dimethylamino)-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₁BrN₂O
Melting Pt: 241…243 °C
MDL Number: MFCD14584844
CAS Number: 1198475-47-6

Order Now

Specification Test Results

Melting point 241-243°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR