5-Amino-3-methyl-1H-pyrazole-4-carboxamide ≥95%
Supplier: Matrix Scientific
054555-1G
054555-500MG
101907-752EA
467.69
USD
101907-752
101907-750
5-Amino-3-methyl-1H-pyrazole-4-carboxamide ≥95%
5-Amino-3-methyl-1H-pyrazole-4-carboxamide
Hazard: Irritant
Formula:
C₅H₈N₄O Melting Pt: 185…187 °C |
MDL Number:
MFCD02020549 CAS Number: 908808-35-5 |
Specification Test Results
Melting point | 185-187°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...