2,5-Dichloroisonicotinamide ≥95%

Supplier: Matrix Scientific

054549-500MG 054549-1G
101907-730EA 556.92 USD
101907-730 101907-732
2,5-Dichloroisonicotinamide ≥95%
2,5-Dichloroisonicotinamide

Hazard: Irritant

Formula: C₆H₄Cl₂N₂O
Melting Pt: 175…178 °C
MDL Number: MFCD14584837
CAS Number: 1221791-56-5

Order Now

Specification Test Results

Melting point 175-178°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR