2-Chloro-6-(chloromethyl)pyridine ≥95%

Supplier: Matrix Scientific

054544-500MG 054544-1G
101907-710EA 556.92 USD
101907-710 101907-712
2-Chloro-6-(chloromethyl)pyridine ≥95%
2-Chloro-6-(chloromethyl)pyridine

Hazard: Irritant

Formula: C₆H₅Cl₂N
Melting Pt: 42…44 °C
MDL Number: MFCD07774109
CAS Number: 78846-88-5

Order Now

Specification Test Results

Melting point 42-44°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR