tert-Butyl-1-thia-4,8-diazaspiro[4.5]decane-8-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

054542-1G 054542-500MG
101907-704EA 467.69 USD
101907-704 101907-702
tert-Butyl-1-thia-4,8-diazaspiro[4.5]decane-8-carboxylate hydrochloride ≥95%
tert-Butyl-1-thia-4,8-diazaspiro[4.5]decane-8-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O₂S
MW: 294.85 g/mol
Melting Pt: 197…199 °C
MDL Number: MFCD14584832
CAS Number: 1221792-10-4

Order Now

Specification Test Results

Melting point 197-199°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR