Ethyl-1-thia-4-azaspiro[4.4]nonane-3-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

054532-500MG
101907-660EA 375.38 USD
101907-660
Ethyl-1-thia-4-azaspiro[4.4]nonane-3-carboxylate hydrochloride ≥95%
Ethyl-1-thia-4-azaspiro[4.4]nonane-3-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClNO₂S
Melting Pt: 159…161 °C
MDL Number: MFCD14584823
CAS Number: 1236272-33-5

Order Now

Specification Test Results

Melting point 159-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR