1-Thia-4-azaspiro[4.5]decane hydrochloride ≥95%

Supplier: Matrix Scientific

054523-500MG 054523-1G
101907-620EA 278.46 USD
101907-620 101907-622
1-Thia-4-azaspiro[4.5]decane hydrochloride ≥95%
1-Thia-4-azaspiro[4.5]decane hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNS
Melting Pt: 171…173 °C
MDL Number: MFCD14584815
CAS Number: 933-41-5

Order Now

Specification Test Results

Melting point 171-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR