3H-Spiro[1,3-benzothiazole-2,1'-cyclohexane] ≥95%
Supplier: Matrix Scientific
054520-1G
054520-500MG
101907-610EA
350.77
USD
101907-610
101907-608
3H-Spiro[1,3-benzothiazole-2,1'-cyclohexane] ≥95%
3H-Spiro[1,3-benzothiazole-2,1'-cyclohexane]
Hazard: Irritant
Formula:
C₁₂H₁₅NS Melting Pt: 111…113 °C |
MDL Number:
MFCD00022809 CAS Number: 182-53-6 |
Specification Test Results
Melting point | 111-113°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...