Ethyl-3-cyano-2,7-dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylate ≥95%
Supplier: Matrix Scientific
054519-1G
054519-5G
054519-500MG
101907-606EA
2129.23
USD
101907-606
101907-602
101907-604
Ethyl-3-cyano-2,7-dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylate ≥95%
Ethyl-3-cyano-2,7-dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₂N₄O₂ Melting Pt: 139…141 °C |
MDL Number:
MFCD01540996 CAS Number: 309938-93-0 |
Specification Test Results
Melting point | 139-141°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...