3-Chloro-4-cyano-5-(trifluoromethyl)-2-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

054518-500MG
101907-600EA 1855.38 USD
101907-600
3-Chloro-4-cyano-5-(trifluoromethyl)-2-pyridine carboxylic acid ≥95%
3-Chloro-4-cyano-5-(trifluoromethyl)-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₈H₂ClF₃N₂O₂
Melting Pt: 122 °C (dec.)
MDL Number: MFCD14584813
CAS Number: 1221791-55-4

Order Now

Specification Test Results

Melting point 122°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR