Ethyl-3-chloro-4-cyano-5-(trifluoromethyl)picolinate ≥95%
Supplier: Matrix Scientific
054516-500MG
054516-1G
101907-594EA
1384.62
USD
101907-594
101907-592
Ethyl-3-chloro-4-cyano-5-(trifluoromethyl)picolinate ≥95%
Ethyl 3-chloro-4-cyano-5-(trifluoromethyl)picolinate
Hazard: Irritant
Formula:
C₁₀H₆ClF₃N₂O₂ Melting Pt: 59…60 °C |
MDL Number:
MFCD14584811 CAS Number: 1221792-27-3 |
Specification Test Results
Melting point | 59-60°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...