Ethyl-3-cyano-6-phenyl-2-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
054514-1G
054514-500MG
101907-586EA
927.69
USD
101907-586
101907-584
Ethyl-3-cyano-6-phenyl-2-pyridinecarboxylate ≥95%
Ethyl-3-cyano-6-phenyl-2-pyridinecarboxylate
Hazard: Irritant
Formula:
C₁₅H₁₂N₂O₂ MW: 252.28 g/mol Melting Pt: 99…101 °C |
MDL Number:
MFCD14584809 CAS Number: 1221792-25-1 |
Specification Test Results
Melting point | 99-101°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...