8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-7-carbonitrile ≥95%
Supplier: Matrix Scientific
054509-500MG
101907-566EA
923.08
USD
101907-566
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-7-carbonitrile ≥95%
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-7-carbonitrile
Hazard: Irritant
Formula:
C₉H₃ClF₃N₃ MW: 245.59 g/mol Melting Pt: 119…120 °C |
MDL Number:
MFCD14584805 CAS Number: 1221792-55-7 |
Specification Test Results
Melting point | 119-120°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...