3,5-Dichloro-2-methoxyisonicotinonitrile ≥95%

Supplier: Matrix Scientific

054506-500MG
101907-554EA 921.54 USD
101907-554
3,5-Dichloro-2-methoxyisonicotinonitrile ≥95%
3,5-Dichloro-2-methoxyisonicotinonitrile

Hazard: Irritant

Formula: C₇H₄Cl₂N₂O
Melting Pt: 91…92 °C
MDL Number: MFCD14584802
CAS Number: 1221791-88-3

Order Now

Specification Test Results

Melting point 91-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR