2,3,5-Trichloroisonicotinamide ≥95%

Supplier: Matrix Scientific

054502-1G 054502-500MG
101907-540EA 927.69 USD
101907-540 101907-538
2,3,5-Trichloroisonicotinamide ≥95%
2,3,5-Trichloroisonicotinamide

Hazard: Irritant

Formula: C₆H₃Cl₃N₂O
Melting Pt: 174…175 °C
MDL Number: MFCD14584798
CAS Number: 70593-53-2

Order Now

Specification Test Results

Melting point 174-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR