2-Chloro-4,6-diiodo-3-methoxypyridine ≥95%

Supplier: Matrix Scientific

054496-500MG 054496-1G
101907-512EA 375.38 USD
101907-512 101907-514
2-Chloro-4,6-diiodo-3-methoxypyridine ≥95%
2-Chloro-4,6-diiodo-3-methoxypyridine

Hazard: Irritant

Formula: C₆H₄ClI₂NO
Melting Pt: 120…122 °C
MDL Number: MFCD14584792
CAS Number: 1221572-27-5

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR