N-[2-Amino-4-bromo-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide ≥95%

Supplier: Matrix Scientific

054494-1G 054494-500MG
101907-506EA 698.46 USD
101907-506 101907-504
N-[2-Amino-4-bromo-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide ≥95%
N-[2-Amino-4-bromo-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide

Hazard: Irritant

Formula: C₉H₅BrF₆N₂O
Melting Pt: 124…126 °C
MDL Number: MFCD14584790
CAS Number: 156425-22-8

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR