5-Chloro-2-hydroxyphenylbenzene carboxylate ≥95%

Supplier: Matrix Scientific

054489-500MG
101907-484EA 556.92 USD
101907-484
5-Chloro-2-hydroxyphenylbenzene carboxylate ≥95%
5-Chloro-2-hydroxyphenylbenzene carboxylate

Hazard: Irritant

Formula: C₁₃H₉ClO₃
MW: 248.67 g/mol
Melting Pt: 140…142 °C
MDL Number: MFCD14584785
CAS Number: 5876-98-2

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR