2-(2-Fluorophenyl)-2-oxoethyl thiocyanate ≥95%

Supplier: Matrix Scientific

047721-500MG 047721-1G
101881-088EA 375.38 USD
101881-088 101881-090
2-(2-Fluorophenyl)-2-oxoethyl thiocyanate ≥95%
2-(2-Fluorophenyl)-2-oxoethyl thiocyanate

Hazard: Irritant

Formula: C₉H₆FNOS
Melting Pt: 53…55 °C
MDL Number: MFCD09027127
CAS Number: 887625-18-5

Order Now

Specification Test Results

Melting point 53-55°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR