N-(4-Fluoro-3-nitrophenyl)-2-(1-pyrrolidinyl)acetamide ≥95%

Supplier: Matrix Scientific

047713-500MG
101881-050EA 181.54 USD
101881-050
N-(4-Fluoro-3-nitrophenyl)-2-(1-pyrrolidinyl)acetamide ≥95%
N-(4-Fluoro-3-nitrophenyl)-2-(1-pyrrolidinyl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₄FN₃O₃
MW: 267.26 g/mol
Melting Pt: 105…106 °C
MDL Number: MFCD09027123
CAS Number: 952182-62-6

Order Now

Specification Test Results

Melting point 105-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR