N-(1,3-Benzodioxol-5-ylmethyl)-3-nitroaniline ≥95%

Supplier: Matrix Scientific

047693-1G 047693-500MG
101881-000EA 181.54 USD
101881-000 101881-002
N-(1,3-Benzodioxol-5-ylmethyl)-3-nitroaniline ≥95%
N-(1,3-Benzodioxol-5-ylmethyl)-3-nitroaniline

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₄
Melting Pt: 99…100 °C
MDL Number: MFCD00422251
CAS Number: 191595-08-1

Order Now

Specification Test Results

Melting point 99-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR