N-(4-Methoxybenzyl)-3-nitroaniline ≥95%

Supplier: Matrix Scientific

047692-1G 047692-500MG
101880-996EA 135.7 USD
101880-996 101880-998
N-(4-Methoxybenzyl)-3-nitroaniline ≥95%
N-(4-Methoxybenzyl)-3-nitroaniline

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₃
Melting Pt: 98…99 °C
MDL Number: MFCD00024365
CAS Number: 305851-22-3

Order Now

Specification Test Results

Melting point 98-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR