Methyl-3,5-dichloro-4-methoxybenzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3,5-dichloro-4-methoxybenzoate
047675-10G
047675-5G
047675-1G
101880-940EA
172.31
USD
101880-940
101880-944
101880-942
Methyl-3,5-dichloro-4-methoxybenzenecarboxylate ≥95%
Methyl-3,5-dichloro-4-methoxybenzenecarboxylate
Hazard: Irritant
Formula:
C₉H₈Cl₂O₃ MW: 235.07 g/mol Melting Pt: 74…77 °C |
MDL Number:
MFCD00017189 CAS Number: 24295-27-0 |
Specification Test Results
Melting point | 74-77°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...