1-(4-Fluoro-3-nitrophenyl)-1H-pyrrole-2,5-dione ≥95%

Supplier: Matrix Scientific

Synonyms: N-(4-Fluoro-3-nitrophenyl)maleimide

047653-500MG 047653-1G
101880-886EA 181.54 USD
101880-886 101880-888
1-(4-Fluoro-3-nitrophenyl)-1H-pyrrole-2,5-dione ≥95%
1-(4-Fluoro-3-nitrophenyl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₀H₅FN₂O₄
MW: 236.16 g/mol
Melting Pt: 127…128 °C
MDL Number: MFCD00591947
CAS Number: 67154-40-9

Order Now

Specification Test Results

Melting point 127-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR