4'-(Naphth-1-ylmethoxy)acetophenone ≥95%

Supplier: Matrix Scientific

047647-500MG 047647-1G
101880-870EA 135.7 USD
101880-870 101880-872
4'-(Naphth-1-ylmethoxy)acetophenone ≥95%
4'-(Naphth-1-ylmethoxy)acetophenone

Hazard: Irritant

Formula: C₁₉H₁₆O₂
Melting Pt: 150…152 °C
MDL Number: MFCD07021293
CAS Number: 885267-47-0

Order Now

Specification Test Results

Melting point 150-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR