1-(4-Hydroxyphenyl)-3-(2,3,4-trimethoxyphenyl)-2-propen-1-one ≥95%
Supplier: Matrix Scientific
047605-1G
047605-500MG
101880-754EA
232.31
USD
101880-754
101880-752
1-(4-Hydroxyphenyl)-3-(2,3,4-trimethoxyphenyl)-2-propen-1-one ≥95%
1-(4-Hydroxyphenyl)-3-(2,3,4-trimethoxyphenyl)-2-propen-1-one
Hazard: Irritant
Formula:
C₁₈H₁₈O₅ Melting Pt: 155…156 °C |
MDL Number:
MFCD03788394 CAS Number: 556028-14-9 |
Specification Test Results
Melting point | 155-156°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...